Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MTDPMMDFF DDANLFGETL EGLSDDAFVQ PGPVSLVDEL NLGAEFEPLH IDSLNHVQGT PTHQKMTDFE QLNQFDSIKF HHVNQSFGSP AEHVLSPHSQ FNCSPIHPQN QPNGLFPDVS DGSPMWGHQT ATTISNQNGS PFHQQGHSHS MHQNKSFVAH HDFALFQANE QQTQCTSLRS QQNRNNLNPG QNSLSQSKNF MNVSGPHRVN VNHPPQMTNA SNSQQSISMQ QFSQTSNPSA HFHKCSSHQE GNFNGPSPNM TSCSVSNSQQ FSSHYSFSSN HISPNSLLQS SAVLASNHTN QTLSDFTGSN SFSPHRGIKQ ESTQHILNPN TSLNSNNFQI LHSSHPQGNY S (1-350 aa encoded by BC033770)
CHD9 (chromodomain-helicase-DNA-binding protein 9), also known as chromatin-related mesenchymal modulator (CReMM), PPAR-α-interacting complex protein, kismet homolog 2 or CHROM1, is a 2,897 amino acid protein belonging to the Snf2/Rad54 helicase family. The CHD family of proteins are ATP-dependent chromatin remodeling enzymes which combine chromodomains with SWI2/Snf2 ATPase/helicase motifs and DNA-binding capability. Localized to the cytoplasm and the nucleus, CHD9 contains two chromodomains, one ATPbinding helicase domain and one C-terminal helicase domain. Chromodomains are protein regions of about 40-50 amino acid residues found in proteins associated with chromatin remodeling and manipulation. The domain is highly conserved among both plants and animals and is found in a large variety of proteins from many genomes. CHD9 acts as a transcriptional coactivator for PPARα and may also be an ATP-dependent chromatin remodeling protein. CHD9 is widely expressed at low levels and is present as three isoforms produced by alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase CHD9; BC022889; CHD-9; chromatin remodeling factor CHROM1; Chromatin-related mesenchymal modulator; Chromatin-remodeling factor CHROM1; Chromodomain-helicase-DNA-binding protein 9; ciprofibrate bound protein p240; CReMM; FLJ12178; Kismet homolog 2; Peroxisomal proliferator-activated receptor A-interacting complex 320 kDa protein; PPAR-alpha-interacting complex protein 320 kDa; PPAR{gamma}-interacting cofactor 320 kDa; proteinx0008
Gene Aliases: AD-013; AD013; CHD-9; CHD9; CReMM; KIAA0308; KISH2; PRIC320; x0008
UniProt ID: (Human) Q3L8U1
Entrez Gene ID: (Human) 80205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support