Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
CHERP is involved in calcium homeostasis, growth and proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium homeostasis endoplasmic reticulum protein; CHERP; ERPROT 213-21; ERPROT213-21; protein with polyglutamine repeat; SR-related CTD associated factor 6; SR-related CTD-associated factor 6
Gene Aliases: 5730408I11Rik; CHERP; D8Wsu96e; DAN16; DAN26; SCAF6; SRA1
UniProt ID: (Human) Q8IWX8, (Mouse) Q8CGZ0
Entrez Gene ID: (Human) 10523, (Mouse) 27967, (Rat) 290614
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support