- Contact Us
- Quick Order
-
Don't have an account ? Create Account
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Human YKL-39/CHI3L2.
CHI3L2 may bind glycan structure with high affinity, but not heparin. Has no chitotriosidase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chitinase 3-like 2; Chitinase-3-like protein 2; Chondrocyte protein 39; YKL-39
Gene Aliases: CHI3L2; CHIL2; YKL-39; YKL39
UniProt ID: (Human) Q15782
Entrez Gene ID: (Human) 1117
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support