Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
CHIA degrades chitin and chitotriose. May participate in the defense against nematodes and other pathogens. There are 3 named isoforms produced by alternative splicing. CHIA is induced via a T helper-2 (Th2)-specific, interleukin-13-mediated pathway in epithelial cells and macrophages. CHIA may be an important mediator of IL13-induced responses in Th2-dominated disorders such as asthma. CHIA hydrolysis N-acetyl-beta-D-glucosaminide 1,4-beta-linkages in chitin and chitodextrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acidic mammalian chitinase; AMCase; Lung-specific protein TSA1902
Gene Aliases: AMCASE; CHIA; CHIT2; TSA1902
UniProt ID: (Human) Q32W80
Entrez Gene ID: (Human) 27159
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support