Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The antibody is specific to CHKA.
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHETK-alpha; Choline kinase alpha; choline kinase R; choline/ethanolamine kinase alpha; CK; EK; Ethanolamine kinase
Gene Aliases: Chetk-alpha; CHK; CHKA; ChoK; CK; CK-R; CK/EK-alpha; CKI; Ckr; EK; EtnK-alpha
UniProt ID: (Human) P35790, (Mouse) Q8BPL3, (Rat) Q01134
Entrez Gene ID: (Human) 1119, (Mouse) 12660, (Rat) 29194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support