Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MADNLPTEFD VVIIGTGLPE SILAAACSRS GQRVLHIDSR SYYGGNWASF SFSGLLSWLK EYQQNNDIGE ESTVVWQDLI HETEEAITLR KKDETIQHTE AFCYASQDME DNVEEIGALQ KNPSLGVSNT FTEVLDSALP EESQLSYFNS DEMPAKHTQK SDTEISLEVT DVEESVEKEK YCGDKTCMHT VSDKDGDKDE SKSTVEDKAD EPIRNRITYS QIVKEGRRFN IDLVSKLLYS QGLLIDLLIK SDVSRYVEFK NVTRILAFRE GKVEQVPCSR ADVFNSKELT MVEKRMLMKF LTFCLEYEQH PDEYQAFRQC SFSEYLKTKK LTPNLQHFVL HSIAMTSESS CTTIDGLNAT KNFLQCLGRF GNTPFLFPLY GQGEIPQGFC RMCAVFGGIY CLRHKVQCFV VDKESGRCKA IIDHFGQRIN AKYFIVEDSY LSEETCSNVQ YKQISRAVLI TDQSILKTDL DQQTSILIVP PAEPGACAVR VTELCSSTMT CMKDTYLVHL TCSSSKTARE DLESVVKKLF TPYTETEINE EELTKPRLLW ALYFNMRDSS GISRSSYNGL PSNVYVCSGP DCGLGNEHAV KQAETLFQEI FPTEEFCPPP PNPEDIIFDG DDKQPEAPGT NNVVMAKLES SEESKNLESP EKHLQN
Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: choroideraemia-like protein; choroideremia-like (Rab escort protein 2); Choroideremia-like protein; Rab escort protein 2; Rab proteins geranylgeranyltransferase component A 2; REP-2
Gene Aliases: CHML; REP2
UniProt ID: (Human) P26374
Entrez Gene ID: (Human) 1122
Molecular Function: G-protein modulator protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support