Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDDTLFQLK FTAKQLEKLA KKAEKDSKAE QAKVKKALLQ KNVECARVYA ENAIRKKNEG VNWLRMASRV DAVASKVQTA VTMKGVTKNM AQVTKALDKA LSTMDLQKVS SVMDRFEQQV QNLDVHTSVM EDSMSSATTL TTPQEQVDSL IMQIAEENGL EVLDQLSQLP EGASAVGESS VRSQEDQLSR RLAALRN (1-196 aa encoded by BC007527)
Probable peripherally associated component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I, -II and -III complexes. ESCRT- III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses). ESCRT-III proteins are believed to mediate the necessary vesicle extrusion and/or membrane fission activities, possibly in conjunction with the AAA ATPase VPS4. Involved in cytokinesis. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells. May also be involved in chromosome condensation. Targets the Polycomb group (PcG) protein BMI1/PCGF4 to regions of condensed chromatin. May play a role in stable cell cycle progression and in PcG gene silencing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: charged multivesicular body protein 1/chromatin modifying protein 1; Charged multivesicular body protein 1a; CHMP1a; chromatin modifying protein 1A; Chromatin-modifying protein 1a; hVps46-1; procollagen (type III) N-endopeptidase; protease, metallo, 1, 33kD; Vacuolar protein sorting-associated protein 46-1; Vps46-1
Gene Aliases: 2900018H07Rik; CHMP1; CHMP1A; KIAA0047; PCH8; PCOLN3; PRSM1; VPS46-1; VPS46A
UniProt ID: (Human) Q9HD42, (Mouse) Q921W0
Entrez Gene ID: (Human) 5119, (Mouse) 234852
Molecular Function:
membrane traffic protein
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support