Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Immunogen sequence: MSNMEKHLF NLKFAAKELS RSAKKCDKEE KAEKAKIKKA IQKGNMEVAR IHAENAIRQK NQAVNFLRMS ARVDAVAARV QTAVTMGKVT KSMAGVVKSM DATLKTMNLE KISALMDKFE HQFETLDVQT QQMEDTMSST TTLTTPQNQV DMLLQEMADE AGLDLNMELP QGQTGSVGTS VASAEQDELS QRLARLRDQV (1-199 aa encoded by BC065933)
CHMP1B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charged multivesicular body protein 1b; charged multivesicular body protein 1b-1; Charged multivesicular body protein 1B1; CHMP1.5; CHMP1b; CHMP1b-1; chromatin modifying protein 1B; Chromatin-modifying protein 1b; Chromatin-modifying protein 1b-1; vacuolar protein sorting 46-2; Vacuolar protein sorting-associated protein 46-2; Vps46-2
Gene Aliases: 2810405I11Rik; C10orf2; C18-ORF2; C18orf2; CHMP1.5; CHMP1B; Chmp1b1; hVps46-2; Vps46-2; Vps46B
UniProt ID: (Human) Q7LBR1, (Mouse) Q99LU0
Entrez Gene ID: (Human) 57132, (Rat) 689364, (Mouse) 67064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support