Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MSVFGKLFGA GGGKAGKGGP TPQEAIQRLR DTEEMLSKKQ EFLEKKIEQE LTAAKKHGTK NKRAALQALK RKKRYEKQLA QIDGTLSTIE FQREALENAN TNTEVLKNMG YAAKAMKAAH DNMDIDKVDE LMQDIADQQE LAEEISTAIS KPVGFGEEFD EDELMAELEE LEQEELDKNL LEISGPETVP LPNVPSIALP SKPAKKKEEE DDDMKELENW AGSM
CHMP4B is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) protein family. The protein is part of the endosomal sorting complex required for transport (ESCRT) complex III (ESCRT-III), which functions in the sorting of endocytosed cell-surface receptors into multivesicular endosomes. The ESCRT machinery also functions in the final abscisson stage of cytokinesis and in the budding of enveloped viruses such as HIV-1. The three proteins of the CHMP4 subfamily interact with programmed cell death 6 interacting protein (PDCD6IP, also known as ALIX), which also functions in the ESCRT pathway. The CHMP4 proteins assemble into membrane-attached 5-nm filaments that form circular scaffolds and promote or stabilize outward budding. These polymers are proposed to help generate the luminal vesicles of multivesicular bodies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charged multivesicular body protein 4b; CHMP4b; chromatin modifying protein 4B; Chromatin-modifying protein 4b; hSnf7-2; hVps32-2; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; SNF7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2
Gene Aliases: C20orf178; CHMP4A; CHMP4B; CTPP3; CTRCT31; dJ553F4.4; SHAX1; SNF7; SNF7-2; Vps32-2; VPS32B
UniProt ID: (Human) Q9H444
Entrez Gene ID: (Human) 128866
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support