Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CHMP4B/C Antibodies

            Bioss

            CHMP4B+CHMP4C Polyclonal Antibody

            View all (2) CHMP4B/C antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # BS-7744R
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            444.00
            100 µL
             
            Estimated availability date: 11-Jun-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CHMP4B+CHMP4C Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            CHMP4B+CHMP4C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            CHMP4B+CHMP4C Antibody (BS-7744R) in IHC (P)

            Formalin-fixed and paraffin embedded rat brain labeled with Anti-CHMP4B/CHMP4C Polyclonal Antibody, Unconjugated (bs-7744R) at 1:200 followed by conjugation to the secondary antibody and DAB staining.
            View Product

            CHMP4B+CHMP4C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CHMP4B+CHMP4C Antibody in Western Blot (WB)
            CHMP4B+CHMP4C Antibody in ELISA (ELISA)

            Product Details

            BS-7744R

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:300
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200
            -

            Immunohistochemistry (Frozen) (IHC (F))

            1:100-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:100-1:500
            -

            ELISA (ELISA)

            1:5,000-1:10,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            KLH conjugated synthetic peptide derived from human CHMP4B/CHMP4C.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

            Contains

            0.02% ProClin 300

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            Probable core component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I,-II and -III complexes. ESCRT-III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses).

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Charged multivesicular body protein 4b; Charged multivesicular body protein 4c; CHMP4b; CHMP4c; chromatin modifying protein 4B; chromatin modifying protein 4C; Chromatin-modifying protein 4b; Chromatin-modifying protein 4c; hSnf7-2; hSnf7-3; hVps32-2; hVps32-3; SNF7 homolog associated with Alix 1; SNF7 homolog associated with Alix 3; Snf7 homologue associated with Alix 1; Snf7 homologue associated with Alix 3; SNF7-2; SNF7-3; Vacuolar protein sorting-associated protein 32-2; Vacuolar protein sorting-associated protein 32-3; vacuolar protein-sorting-associated protein 32-2; Vps32-2; Vps32-3

            View more View less

            Gene Aliases: 2010012F05Rik; 2010012P02Rik; 2210015K02Rik; 2310010I16Rik; C20orf178; C76846; CHMP4A; CHMP4B; CHMP4C; CTPP3; CTRCT31; dJ553F4.4; RGD1309846; RGD1565889; SHAX1; SHAX3; SNF7; SNF7-2; SNF7-3; Vps32-2; VPS32B; VPS32C

            View more View less

            UniProt ID: (Human) Q9H444, (Human) Q96CF2, (Mouse) Q9D8B3, (Mouse) Q9D7F7, (Rat) Q569C1

            View more View less

            Entrez Gene ID: (Human) 128866, (Human) 92421, (Mouse) 75608, (Rat) 100359642, (Mouse) 66371, (Rat) 361916

            View more View less

            Function(s)

            protein binding identical protein binding protein homodimerization activity cadherin binding involved in cell-cell adhesion membrane traffic protein

            Process(es)

            mitotic cytokinesis cell separation after cytokinesis posttranslational protein targeting to membrane autophagy nucleus organization mitotic metaphase plate congression exit from mitosis regulation of centrosome duplication endosomal transport viral life cycle nuclear envelope reassembly multivesicular body assembly maintenance of lens transparency viral budding via host ESCRT complex viral budding regulation of viral process protein homooligomerization negative regulation of cell death membrane fission ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway cell-cell adhesion negative regulation of neuron death regulation of mitotic spindle assembly positive regulation of viral release from host cell negative regulation of autophagosome assembly abscission protein transport negative regulation of cytokinesis transport vacuolar transport bioluminescence
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-rms4r:80/100.66.77.126:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0