Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QTRNFRLYYD GKHFVGEKRF ESIHDLVTDG LITLYIETKA AEYIAKMTIN PIYEHVGYTT LNREPAYKKH MPVLKETHDE RDSTGQDGVS EKRLTSLVRR ATLKENEQIP
GTPase-activating protein for p21-rac and a phorbol ester receptor. Involved in the assembly of neuronal locomotor circuits as a direct effector of EPHA4 in axon guidance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-chimaerin; a2-chimaerin; Alpha-chimerin; chimaerin 1; chimerin (chimaerin) 1; N-chimaerin; N-chimerin; NC; Rho GTPase-activating protein 2
Gene Aliases: ARHGAP2; CHN; CHN1; DURS2; NC; RHOGAP2
UniProt ID: (Human) P15882
Entrez Gene ID: (Human) 1123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support