Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the final step of de novo phosphatidylcholine (PC) synthesis, i.e. the transfer of choline phosphate from CDP-choline to the free hydroxyl of a diacylglycerol (DAG), producing a PC. It thereby plays a central role in the formation and maintenance of vesicular membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAPT1-like protein; Cholinephosphotransferase 1; cholinephosphotransferase 1 alpha; Diabetic nephropathy-associated gene transcript 1; Diacylglycerol cholinephosphotransferase 1; hCPT1; mCHPT1; phosphatidylcholine synthesizing enzyme
Gene Aliases: CHPT1; CPT; CPT1; Dn4; MSTP022
UniProt ID: (Human) Q8WUD6, (Mouse) Q8C025
Entrez Gene ID: (Human) 56994, (Mouse) 212862, (Rat) 362866
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support