Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: MPVYESVFME DGETTRKIAL ETERPPQVEV HVWTIRKGIL QHFHIEKISK RMFEELPHFK LVTRTTLSQW KIFTEGEAQI SQMCSSRVCR TELEDLVKVL YLERSEKGHC
CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chordin-like 1; Chordin-like protein 1; neuralin 1; Neuralin-1; Neurogenesin-1; Ventroptin
Gene Aliases: CHL; CHRDL1; dA141H5.1; MGC1; MGCN; NRLN1; VOPT
UniProt ID: (Human) D3DUY6
Entrez Gene ID: (Human) 91851
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support