Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
After binding acetylcholine, the ACHR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor; cholinergic receptor, nicotinic alpha 2; cholinergic receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal); Neuronal acetylcholine receptor subunit alpha-2
Gene Aliases: CHRNA2
UniProt ID: (Human) Q15822
Entrez Gene ID: (Human) 1135
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support