Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31683 has been tested in indirect ELISA.
After binding acetylcholine, the ACHR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor; cholinergic receptor, nicotinic alpha 2; cholinergic receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal); Neuronal acetylcholine receptor subunit alpha-2; nicotinic acetylcholine receptor
Gene Aliases: ACHR; Acra2; CHRNA2
UniProt ID: (Human) Q15822, (Rat) P12389
Entrez Gene ID: (Human) 1135, (Rat) 170945
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support