Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31698 has been tested in indirect ELISA.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor epsilon; Acetylcholine receptor subunit epsilon; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; ACHRG; cholinergic receptor; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; epsilon-subunit; gene for acetylcholine receptor epsilon subunit; MGC133376
Gene Aliases: ACHRE; CHRNE; CMS1D; CMS1E; CMS2A; CMS4A; CMS4B; CMS4C; FCCMS; SCCMS
UniProt ID: (Human) D3DTK6, (Rat) P09660
Entrez Gene ID: (Human) 1145, (Rat) 29422
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support