Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CHST11.
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Can also sulfate Gal residues in desulfated dermatan sulfate. Preferentially sulfates in GlcA->GalNAc unit than in IdoA->GalNAc unit. Does not form 4, 6- di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C4S-1; C4ST; C4ST-1; Carbohydrate sulfotransferase 11; chondroitin 4; Chondroitin 4-O-sulfotransferase 1; Chondroitin 4-sulfotransferase 1; IgH/CHST11 fusion
Gene Aliases: 1110020P09Rik; C4s; C4ST; C4ST-1; C4ST1; CHST11; HSA269537
UniProt ID: (Human) Q9NPF2, (Mouse) Q9JME2, (Rat) P69478
Entrez Gene ID: (Human) 50515, (Mouse) 58250, (Rat) 314694
Molecular Function: transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support