Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CHST12.
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C4ST-2; Carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; Chondroitin 4-sulfotransferase 2; Sulfotransferase Hlo
Gene Aliases: AI595374; C4S-2; C4ST-2; C4ST2; CHST12; UNQ500/PRO1017
UniProt ID: (Human) Q9NRB3, (Mouse) Q99LL3
Entrez Gene ID: (Human) 55501, (Mouse) 59031, (Rat) 304322
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support