Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: RDRELTADSD VDEFLDKFLS AGVKQSDLPR KETEQPPAPG SMEESVRGYD WSPRDARRSP DQGRQQAERR SVLRGFCANS SLAFPTKERA FDDIP
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C4ST-2; Carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; Chondroitin 4-sulfotransferase 2; Sulfotransferase Hlo
Gene Aliases: C4S-2; C4ST-2; C4ST2; CHST12; UNQ500/PRO1017
UniProt ID: (Human) Q9NRB3
Entrez Gene ID: (Human) 55501
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support