Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CHST3 is an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrixand most cells which is involved in cell migration and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C6ST; C6ST-1; carbohydrate (chondroitin 6) sulfotransferase 3; Carbohydrate sulfotransferase 3; chondroitin 6 sulfotransferase 1; Chondroitin 6-O-sulfotransferase 1; Chondroitin 6-sulfotransferase; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0; GST-0
Gene Aliases: C6ST; C6ST-1; C6ST1; CHST3; GST-0; Gst0; HSD
UniProt ID: (Human) Q7LGC8, (Mouse) O88199
Entrez Gene ID: (Human) 9469, (Mouse) 53374
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support