Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NKFSNMNWP VDIHPLNKSL VKDNKWKKTE ETQEKRRSFL QEFCKKYGGV SHHQSHLFHT VSRIYVEDKH KILYCEVPKA GCSNWKRILM VLNGLASSAY NISHNAVHYG KHLKKLDSFD LKGIYTRLNT YTKAVFVRDP MERLVSAFRD KFEHPNSYYH PVFGKAIIKK YRPNACEEAL INGSGVKFKE FIHYLLDSHR PVGMDIHWEK VSKLCYPCLI NYDFVGKFET LEEDANYFLQ MIGAPKELKF PNFKDRHSSD ERTNAQVVRQ YLKDLTRTER QLIYDFYYLD YLMFNYTTPF L (144-443 aa encoded by BC025764)
Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST9, carry out sulfation of carbohydrates (Hiraoka et al., 2001 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; Carbohydrate sulfotransferase 9; GalNAc-4-O-sulfotransferase 2; GalNAc-4-ST2; GalNAc-4-sulfotransferase 2; GalNAc4ST-2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2
Gene Aliases: 5430438D01Rik; CHST9; GALNAC4ST-2; GalNAc4ST2; UNQ2549/PRO6175
UniProt ID: (Human) Q7L1S5, (Mouse) Q76EC5
Entrez Gene ID: (Human) 83539, (Mouse) 71367, (Rat) 291770
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support