Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: Jurkat, Raji, K562, THP-1, HL-60, NCI-H929, NIH-3T3, JC, BCL-1.
Predicted reactivity: Mouse (81%), Rat (81%), Chimpanzee (99%), Bovine (90%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anamorsin; anamorsin {ECO:0000255|HAMAP-Rule:MF_03115}; Cytokine-induced apoptosis inhibitor 1; cytokine-induced apoptosis inhibitor 1 {ECO:0000255|HAMAP-Rule:MF_03115}; Fe-S cluster assembly protein DRE2 homolog; fe-S cluster assembly protein DRE2 homolog {ECO:0000255|HAMAP-Rule:MF_03115}; predicted protein of HQ0915
Gene Aliases: 2810413N20Rik; AA617265; Anamorsin; AU021794; CIAPIN1; CUA001; DRE2; PRO0915
UniProt ID: (Human) Q6FI81, (Rat) Q5XID1, (Mouse) Q8WTY4
Entrez Gene ID: (Human) 57019, (Rat) 307649, (Mouse) 109006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support