Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MRGDRASGGP GNHNGSWARE GPRLGPSWKR GLWSPRGGPN RPAEPSRPLT FMGSQTKRVL FTPLMHPARP FRVSNHDRSS RRGVMASSLQ ELISKTLDAL VIATGLVTLV LEEDGTVVDT EEFFQTLGDN THFMILEKGQ KWMPGSQHVP TCSPPKRSGI ARVTFDLYRL NPKDFIGCLN VKATMYEMYS VSYDIRCTGL KGLLRSLLRF LSYSAQVTGQ FLIYLGTYML RVLDDKEERP SLRSQAKGRF TCG
This gene encodes the homolog of the mouse protein Cidea that has been shown to activate apoptosis. This activation of apoptosis is inhibited by the DNA fragmentation factor DFF45 but not by caspase inhibitors. Mice that lack functional Cidea have higher metabolic rates, higher lipolysis in brown adipose tissue and higher core body temperatures when subjected to cold. These mice are also resistant to diet-induced obesity and diabetes. This suggests that in mice this gene product plays a role in thermogenesis and lipolysis. Alternatively spliced transcripts have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell death activator CIDE-A; Cell death-inducing DFFA-like effector A; cide a; Lipid transferase CIDEA
Gene Aliases: CIDE-A; CIDEA
UniProt ID: (Human) O60543
Entrez Gene ID: (Human) 1149
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support