Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blot applications, this antibody detects a band at ~25kDa. A suggested positive control is mouse liver tissue lysate.
PA5-19909 can be used with blocking peptide PEP-0035.
CIDEB activates apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell death activator CIDE-B; Cell death-inducing DFFA-like effector B; CID; DFFA-like B; Lipid transferase CIDEB
Gene Aliases: 1110030C18Rik; AI790179; CIDE-B; Cideb
UniProt ID: (Mouse) O70303
Entrez Gene ID: (Mouse) 12684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support