Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
CIPC (CLOCK Interacting Pacemaker) is associated with Ectrodactyly, Ectodermal Dysplasia, and Cleft Lip/Palate Syndrome 1. Among its related pathways are Circadian rythm related genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLOCK-interacting circadian protein; CLOCK-interacting pacemaker; CLOCK-interacting protein, circadian
Gene Aliases: 2310044G17Rik; AI842544; AI853744; CIPC; KIAA1737; mKIAA1737
UniProt ID: (Human) Q9C0C6, (Mouse) Q8R0W1
Entrez Gene ID: (Human) 85457, (Mouse) 217732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support