Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31702 has been tested in indirect ELISA.
CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A18 hnRNP; cold inducible RNA-binding protein; Cold-inducible RNA-binding protein; glycine-rich RNA binding protein; Glycine-rich RNA-binding protein CIRP; testicular tissue protein Li 39
Gene Aliases: A18HNRNP; CIRBP; CIRP
UniProt ID: (Human) B4E2X2
Entrez Gene ID: (Human) 1153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support