Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: NWLAASGTSA GVHVYNVKQL KLHCTVPAYN FPVTAMAIAP NTNNLVIAHS DQQVFEYSIP DKQYTDWSRT VQKQGFHHLW LQRDTPITHI SFHPKRPMHI LLHDAYMFCI IDKSLPLPND KTLLYNPFPP TNESDVIRRR TAHAFKISKI YKPLLFMDLL DERTLVAVER PLDDIIAQLP PPIKKKKFGT
CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cirhin; cirrhosis, autosomal recessive 1A (cirhin); testis expressed gene 292; Testis-expressed gene 292 protein; U3 small nucleolar RNA-associated protein 4 homolog; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog
Gene Aliases: CIRH1A; CIRHIN; cPERP-E; KIAA1988; NAIC; Teg-292; TEX292; UTP4
UniProt ID: (Mouse) Q8R2N2, (Human) Q969X6
Entrez Gene ID: (Mouse) 21771, (Human) 84916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support