Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The antibody recognizes all the isoforms of CIT.
This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: citron (rho-interacting, serine/threonine kinase 21); citron kinase; Citron Rho-interacting kinase; citron-K kinase; citron-N; CRIK; CTRO; postsynaptic density protein (citron); Rho-interacting, serine/threonine-protein kinase 21; serine/threonine kinase 21; Serine/threonine-protein kinase 21
Gene Aliases: C030025P15Rik; CIT; Cit-k; CRIK; CRIK-SK; KIAA0949; STK21
UniProt ID: (Human) O14578, (Mouse) P49025, (Rat) E9PSL7
Entrez Gene ID: (Human) 11113, (Mouse) 12704, (Rat) 83620
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support