Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
There are two isoforms of CITED2 protein and the calculated molecular weight of them are 24kDa and 28 kDa.
Immunogen sequence: MADHMMAMN HGRFPDGTNG LHHHPAHRMG MGQFPSPHHH QQQQPQHAFN ALMGEHIHYG AGNMNATSGI RHAMGPGTVN GGHPPSALAP AARFNNSQFM GPPVASQGGS LPASMQLQKL NNQYFNHHPY PHNHYMPDLH PAAGHQMNGT NQHFRDCNPK HSGGSSTPGG SGGSSTPGGS GSSSGGGAGS SNSGGGSGSG NMPASVAHVP AAMLPPNVID TDFIDEEVLM SLVIEMGLDR IKELPELWLG QNEFDFMTDF VCKQQPSRVS C (1-270 aa encoded by BC004377)
The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; MRG-1; MSG-related protein 1; MSG1-related gene 1; P35srj
Gene Aliases: AI835299; ASD8; CITED2; ER154-like; MRG-1; MRG1; Msg2; P35SRJ; VSD2
UniProt ID: (Human) Q5VTF4, (Mouse) O35740
Entrez Gene ID: (Human) 10370, (Rat) 114490, (Mouse) 17684
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support