Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSHGAGLVRT TCSSGSALGP GAGAAQPSAS PLEGLLDLSY PRTHAALLKV AQMVTLLIAF ICVRSSLWTN YSAYSYFEVV TICDLIMILA FYLVHLFRFY RVLTCISWPL SELLHYLIGT LLLLIASIVA ASKSYNQSGL VAGAIFGFMA TFLCMASIWL SYKISCVTQS TDAAV
CMTM7 belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. The protein encoded by this gene is highly expressed in leukocytes, but its exact function is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chemokine-like factor super family 7; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; Chemokine-like factor superfamily member 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7
Gene Aliases: CKLFSF7; CMTM7
UniProt ID: (Human) Q96FZ5
Entrez Gene ID: (Human) 112616
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support