Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to CKM.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CPK-M; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, muscle; Creatine phosphokinase M-type; M-CK
Gene Aliases: CKM; CKMM; M-CK; MCK
UniProt ID: (Human) P06732, (Rat) P00564, (Mouse) P07310
Entrez Gene ID: (Human) 1158, (Rat) 24265, (Mouse) 12715
Molecular Function:
amino acid kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support