Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 93%
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chloride channel 5; Chloride channel protein 5; Chloride transporter ClC-5; ClC-5; H(+)/Cl(-) exchange transporter 5; hClC-K2
Gene Aliases: 5430408K11Rik; ClC-5; CLC5; Clcn5; D930009B12Rik; DXImx42e; Sfc13; T25545
UniProt ID: (Rat) P51796, (Mouse) Q9WVD4
Entrez Gene ID: (Rat) 25749, (Mouse) 12728
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support