Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: WVGMVHFDST ATIVNKLIQI KSSDERNTLM AGLPTYPLGG TSICSGIKYA FQVIGELHSQ LDGSEVLLLT DGEDNTASSC IDEV
Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 58%.
The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Alternative splicing results in multiple transcript variants, only one of which is thought to be protein coding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaCC-2; Calcium-activated chloride channel family member 4; Calcium-activated chloride channel protein 2; Calcium-activated chloride channel regulator 4; Chloride channel accessory 4; chloride channel regulator 4; chloride channel, calcium activated, family member 4; hCaCC-2; hCLCA4
Gene Aliases: CaCC; CaCC2; CLCA4; UNQ562/PRO1124
UniProt ID: (Human) Q14CN2
Entrez Gene ID: (Human) 22802
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support