Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chloride channel K2; Chloride channel Kb; Chloride channel protein ClC-Kb; chloride channel, kidney, B; chloride channel, voltage-sensitive Kb; ClC-K2; CLCNK; hClC-Kb; MGC24087; RP11-5P18.8
Gene Aliases: ClC-K2; ClC-K2L; ClC-Kb; Clck2; CLCKB; Clcnk1l; CLCNKB
UniProt ID: (Human) P51801, (Rat) P51802
Entrez Gene ID: (Human) 1188, (Rat) 79430
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support