Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).^M
^M
Immunogen sequence: VTLRTDFSNF TSNTVAEIQA LTSQGSSLEE TIASLKAEVE GFKQERQAVH SEMLLRVQQL VQDLKKLTCQ VATLNNNGEE ASTEGTCCPV NWVEHQDSCY
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; CD301; Macrophage lectin 2; macrophage lectin 2 (calcium dependent)
Gene Aliases: CD301; CLEC10A; CLECSF13; CLECSF14; HML; HML2; MGL
UniProt ID: (Human) Q8IUN9
Entrez Gene ID: (Human) 10462
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support