Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRPAFALCLL WQALWPGPGG GEHPTADRAG CSASGACYSL HHATMKRQAA EEACILRGGA LSTVRAGAEL RAVLALLRAG PGPGGGSKDL LFWVALERRR SHCTLENEPL RGFSWLSSDP GGLESDTLQW VEEPQRSCTA RRCAVLQATG GVEPAGWKEM RCHLRANGYL CKYQFEVLCP APRPGAASNL SYRAPFQLHS AALDFSPPGT EVSALCRGQL PISVTCIADE IGARWDKLSG DVLCPCPGRY LRAGKCAELP NCLDDLGGFA CECATGFELG KDGRSCVTSG EGQPTLGGTG VPTRRPPATA TSPVPQRTWP IRVDEKLGET PLVPEQDNSV TSIPEIPRWG SQSTMSTLQM SLQAESKATI TPSGSVISKF NSTTSSATPQ AFDSSSAVVF IFVSTAVVVL VILTMTVLGL VKLCFHESPS SQPRKESMGP PGLESDPEPA ALGSSSAHCT NNGVKVGDCD LRDRAEGALL AESPLGSSDA
CLEC-14A (C-type lectin domain family 14 member A), also known as EGFR-5 (epidermal growth factor receptor 5) or CEG1, is a 490 amino acid single-pass type I membrane protein that contains one C-type lectin domain and an EGF-like domain. The gene encoding CLEC-14A maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 14 member A; C-type lectin domain family 14, member A; ClECT and EGF-like domain containing protein; EGFR-5; Epidermal growth factor receptor 5
Gene Aliases: C14orf27; CEG1; CLEC14A; EGFR-5; EGFR5; UNQ236/PRO269
UniProt ID: (Human) Q86T13
Entrez Gene ID: (Human) 161198
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support