Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SKASTERAAL LDGHDLLRTN ASKQTAALGA LKEEVGDCHS CCSGTQAQLQ TTRAELGEAQ AKLMEQESAL RELRERVTQG LAEAGRGRED VRTELFRALE AVRLQNNSCE PCPTSWLSFE GSCYFFSVPK TTWAAAQDHC ADASAHLVIV GGLDEQGFLT RNTRGRGYWL GLRAVRHLGK VQGYQWVDGV SLSFSHWNQG EPNDAWGREN CVMMLHTGLW NDAPCDSEKD GWICEKRHNC; Positive Samples: LO2, U-937; Cellular Location: Cell membrane, Single-pass type II membrane protein
This gene encodes a glycan-binding receptor and member of the C-type lectin family which plays a role in the T-cell immune response. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 4 member G; C-type lectin domain family 4, member G; C-type lectin superfamily 4, member G; Liver and lymph node sinusoidal endothelial cell C-type lectin; LSECtin
Gene Aliases: CLEC4G; DTTR431; LP2698; LSECtin; UNQ431; UNQ431/PRO792
UniProt ID: (Human) Q6UXB4
Entrez Gene ID: (Human) 339390
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support