Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: clathrin interacting protein localized in the trans-Golgi region; Clathrin interactor 1; Clathrin-interacting protein localized in the trans-Golgi region; Clint; Enthoprotin; epsin 4; Epsin-4; Epsin-related protein; EpsinR
Gene Aliases: CLINT; CLINT1; ENTH; EPN4; EPNR; KIAA0171
UniProt ID: (Human) Q14677
Entrez Gene ID: (Human) 9685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support