Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVVVYFAEY FINQGLFELL FFWNTSLSHA QQYRWYQMLY QAGVFASRSS LRCCRIRFTW ALALLQCLNL VFLLADVWFG FL (288-368 aa encoded by BC002394)
CLN3 is a highly glycosylated, hydrophobic, 438-amino acid protein with 6 transmembrane domains. The CLN3 protein localizes to the lysosomal membrane and plays a role in lysosomal function. It may act as a chaperone involved in the folding and unfolding of other proteins, namely subunit C of the ATP synthase complex. Mutations in the CLN3 gene cause Batten disease, a recessively inherited neurodegenerative disorder of childhood caused by lysosomal accumulation of hydrophobic material, mainly ATP synthase subunit C. Batten disease is the most common form of a group of disorders known as neuronal ceroid lipofuscinoses (NCLs). Symptoms of Batten disease include progressive loss of vision, seizures, and psychomotor disturbances.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Batten disease protein; Battenin; ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease); ceroid-lipofuscinosis, neuronal 3; CLN 3; Cln3p; MGC102840; Protein CLN3
Gene Aliases: AI323623; BTS; CLN3; JNCL
UniProt ID: (Human) Q13286, (Mouse) Q61124
Entrez Gene ID: (Human) 1201, (Rat) 293485, (Mouse) 12752
Molecular Function: membrane trafficking regulatory protein membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support