Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Immunogen sequence: IPSRRHWPVP YKRFDFRPKP DPYCQAKYTF CPTGSPIPVM EGDDDIEVFR LQAPVWEFKY GDLLGHLKIM HDAIGFRSTL TGKNYTMEWY ELFQLGNCTF PHLRPEMDAP FWCNQGAACF FEGIDDVHWK ENGTLVQVAT ISGNMFNQMA KWVKQDNETG IYYETWNVKA SPEKGAETWF DSYDCSKFVL RTFNKLAEFG AEFKNIETNY TRIFLYSGEP TYLGNETSVF GPTGNKTLGL AIKRFYYPFK PHLPTKEFLL SLLQIFDAVI VHKQFYLFYN FEYWFLPMKF PFIKITYEEI PLPIRNKTLS GL; Positive Samples: Mouse heart, Rat heart; Cellular Location: Lysosome
This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function.[provided by RefSeq, Oct 2008].
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bis(monoacylglycero)phosphate synthase CLN5; BMP synthase CLN5; ceroid-lipofuscinosis; Ceroid-lipofuscinosis neuronal protein 5; ceroid-lipofuscinosis neuronal protein 5 homolog; Palmitoyl protein thioesterase CLN5; Protein CLN5; S-depalmitoylase CLN5
Gene Aliases: A730075N08Rik; BMPS; CLN5; NCL
UniProt ID: (Human) O75503, (Mouse) Q3UMW8
Entrez Gene ID: (Human) 1203, (Rat) 306128, (Mouse) 211286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support