Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: PLIPIVVEQT GRGERAYDIY SRLLRERIVC VMGPIDDSVA SLVIAQLLFL QSESNKKPIH MYINSPGGVV TAGLAIYDTM QYILNPICTW CVGQAASMGS LLLAAGTPGM RHSLPNSRIM IHQPSGGARG QATDIAIQAE EIMKLKKQLY NIYAKHTKQS LQVIESAMER DRYMSPMEAQ EFGILDKVLV HPPQDGEDEP TLVQKEPVEA APAAEPVPAS T
Positive Samples: LO2, A-549, BxPC-3, 22Rv1, Mouse liver, Mouse kidney, Mouse heart, Mouse lung, Rat liver; Cellular Location: Mitochondrion matrix
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Endopeptidase Clp; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Aliases: AU019820; CLPP; D17Wsu160e; DFNB81; PRLTS3
UniProt ID: (Human) Q16740, (Mouse) O88696
Entrez Gene ID: (Human) 8192, (Rat) 301117, (Mouse) 53895
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support