Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MWPGILVGG ARVASCRYPA LGPRLAAHFP AQRPPQRTLQ NGLALQRCLH ATATRALPLI PIVVEQTGRG ERAYDIYSRL LRERIVCVMG PIDDSVASLV IAQLLFLQSE SNKKPIHMYI NSPGGVVTAG LAIYDTMQYI LNPICTWCVG QAASMGSLLL AAGTPGMRHS LPNSRIMIHQ PSGGARGQAT DIAIQAEEIM KLKKQLYNIY AKHTKQSLQV IESAMERDRY MSPMEAQEFG ILDKVLVHPP QDGEDEPTLV QKEPVEAAPA AEPVPAST (1-277 aa encoded by BC002956)
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Endopeptidase Clp; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Aliases: CLPP; DFNB81; PRLTS3
UniProt ID: (Human) Q16740
Entrez Gene ID: (Human) 8192, (Pig) 100737271
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support