Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSFRDLRNF TEMMRALGYP RHISMENFRT PNFGLVSEVL LWLVKRYEPQ TDIPPDVDTE QDRVFFIKAI AQFMATKAHI KLNTKKLYQA DGYAVKELLK ITSVLYNAMK TKGMEGSEIV EEDVNKFKFD LGSKIADLKA ARQLASEITS KGASLYDLLG MEVELREMRT EAIARPLEIN ETEKVMRIAI KEILTQVQKT KDLLNNVASD EANLEAKIEK RKLELERNRK RLETLQSVRP CFMDEYEKTE EELQKQYDTY LEKFQNLTYL EQQLEDHHRM EQERFEEAKN TLCLIQNKLK EEEKRLLKSG SNDDSDIDIQ EDDESDSELE ERRLPKPQTA MEMLMQGRPG KRIVGTMQGG DSDDNEDSEE SEIDMEDDDD EDDDLEDESI SLSPTKPNRR VWKSEPLDES DNDF (1-413 aa encoded by BC017070)
CLUAP1 may play a role in cell proliferation or apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 22; Clusterin-associated protein 1; flagellar associated protein 22, qilin-like protein, homolog; Qilin
Gene Aliases: 2310030D15Rik; 2610111M03Rik; CFAP22; CLUAP1; FAP22; IFT38; KIAA0643
UniProt ID: (Human) Q96AJ1, (Rat) Q6AYJ5, (Mouse) Q8R3P7
Entrez Gene ID: (Human) 23059, (Dog) 479868, (Rat) 363544, (Mouse) 76779
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support