Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CLUAP1 may play a role in cell proliferation or apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 22; Clusterin-associated protein 1; flagellar associated protein 22, qilin-like protein, homolog; Qilin
Gene Aliases: 2310030D15Rik; 2610111M03Rik; CFAP22; CLUAP1; FAP22; IFT38; KIAA0643
UniProt ID: (Human) Q96AJ1, (Rat) Q6AYJ5, (Mouse) Q8R3P7
Entrez Gene ID: (Human) 23059, (Rat) 363544, (Mouse) 76779
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support