Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MRTEAIARPL EINETEKVMR IAIKEILTQV QKTKDLLNNV ASDEANLEAK IEKRKLELER NRKRLETLQS VRPCFMDEYE KTEEELQKQY DTYLEKFQNL TYLEQQLEDH HRMEQERFEE AKNTLCLIQN KLKEEEKRLL KSGSNDDSDI DIQEDDESDS ELEERRLPKP QTAMEMLMQG RPGKRIVGTM QGGDSDDNED SEESEIDMED DDDEDDDLED ESISLSPTKP NRRVRKSEPL DESDNDF
CLUAP1 may play a role in cell proliferation or apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 22; Clusterin-associated protein 1; flagellar associated protein 22, qilin-like protein, homolog; Qilin
Gene Aliases: CFAP22; CLUAP1; FAP22; IFT38; KIAA0643
UniProt ID: (Human) Q96AJ1
Entrez Gene ID: (Human) 23059
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support