Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EGYDSVFSVV RRHQFRWSEI QKGVREVTEP LNLNPAKRPR RQDWDGELYE NGSFYFAKRH LIEMGYLQGG KMAYYEMRAE HSVDIDVDID WPIAEQRVLR
Catalyzes the activation of N-acetylneuraminic acid (NeuNAc) to cytidine 5'-monophosphate N-acetylneuraminic acid (CMP-NeuNAc), a substrate required for the addition of sialic acid. Has some activity toward NeuNAc, N-glycolylneuraminic acid (Neu5Gc) or 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CMP-N-acetylneuraminic acid synthase; CMP-N-acetylneuraminic acid synthetase; CMP-Neu5Ac synthase; CMP-Neu5Ac synthetase; CMP-NeuNAc synthase; CMP-NeuNAc synthetase; CMP-sialic acid synthetase; cytidine 5'-monophosphate N-acetylneuraminic acid synthetase; cytidine monophospho-N-acetylneuraminic acid synthetase; N-acylneuraminate cytidylyltransferase
Gene Aliases: AW208911; CMAS; CMPNeu5Ac; CSS; D6Bwg0250e
UniProt ID: (Human) Q8NFW8, (Mouse) Q99KK2
Entrez Gene ID: (Human) 55907, (Rat) 312826, (Mouse) 12764
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support