Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidylate kinase; cytidylate kinase 1; dCMP kinase; Deoxycytidylate kinase; EC 2.7.4.14; KCY; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); Nucleoside-diphosphate kinase; RP11-511I2.1; ump-cmp; UMP-CMP kinase; UMP/CMP kinase; UMP/CMPK; uridine monophosphate kinase; Uridine monophosphate/cytidine monophosphate kinase
Gene Aliases: 0610011D08Rik; CK; CMK; CMPK; CMPK1; UCK; UMK; UMP-CMPK; UMPK
UniProt ID: (Human) P30085, (Mouse) Q9DBP5
Entrez Gene ID: (Human) 51727, (Dog) 610291, (Mouse) 66588
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support