Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen Sequence: LILIVYCTPF YERVDTTKVK SSDFYITLGT GCVFLLASII FVSTHDRTSA EIAAIVFGFI ASFMFLLDFI TMLYEKRQES QLRKPENTTR AEALTEPLNA
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. Han W., Genomics 81:609-617(2003). Ota T., Nat. Genet. 36:40-45(2004). The MGC Project Team; Genome Res. 14:2121-2127(2004).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chemokine-like factor super family 6; chemokine-like factor superfamily 6; Chemokine-like factor superfamily member 6; CKLF-like MARVEL transmembrane domain containing 6; CKLF-like MARVEL transmembrane domain-containing protein 6
Gene Aliases: 2810051A14Rik; AA536733; CKLFSF6; CMTM6; PRO2219
UniProt ID: (Human) Q9NX76, (Mouse) Q9CZ69
Entrez Gene ID: (Human) 54918, (Mouse) 67213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support