Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Involved in the transport and maturation of proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CNIH-2; Cornichon family AMPA receptor auxiliary protein 2; cornichon homolog 2; cornichon homolog 3; Cornichon-like protein; Protein cornichon homolog 2
Gene Aliases: CNIH-2; CNIH2; Cnih3; CNIL; RGD1304930
UniProt ID: (Human) Q6PI25, (Rat) Q5BJU5, (Mouse) O35089
Entrez Gene ID: (Human) 254263, (Rat) 361705, (Mouse) 12794
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support