Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TIETLMRINA HSRGNAPEGL PQLMEVVRSN YEAMIDRAHG GPNFMMHSGI SQASEYDDPP GLREKAEYLL REWVNLYHSA AAGRDSTKAF SAFVG
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3'UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenal gland protein AD-005; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 1; CCR4-NOT transcription complex, subunit 1; Negative regulator of transcription subunit 1 homolog; NOT1 (negative regulator of transcription 1, yeast) homolog; NOT1H
Gene Aliases: AD-005; CDC39; CNOT1; KIAA1007; NOT1; NOT1H
UniProt ID: (Human) A5YKK6
Entrez Gene ID: (Human) 23019
Molecular Function:
mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support